The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013781.3 | NP_038809.1 | 1 - 2 | 141 | 84 |
NR_045535.1 | 1 - 2 | 141 | 84 | ||
GenBank mRNA | AB043953.1 | - | 1 - 2 | 124 | 84 |
AF168364.1 | - | 1 - 2 | 82 | 84 | |
AK155165.1 | - | 1 - 2 | 77 | 84 | |
AK172072.1 | - | 1 - 2 | 62 | 84 | |
BC113203.1 | - | 1 - 2 | 96 | 84 |