The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252547.1 | NP_001239476.1 | 6 - 7 | 1400 | 63 |
NM_013781.3 | NP_038809.1 | 7 - 8 | 1886 | 63 | |
NR_045535.1 | 8 - 9 | 2344 | 63 | ||
GenBank mRNA | AB030442.1 | - | 6 - 7 | 1363 | 63 |
AB043953.1 | - | 7 - 8 | 1869 | 63 | |
AF168364.1 | - | 7 - 8 | 1827 | 63 | |
AK042709.1 | - | 6 - 7 | 1384 | 63 | |
AK085885.1 | - | - | 1384 | 63 | |
AK155165.1 | - | 7 - 8 | 1822 | 63 | |
AK172072.1 | - | 8 - 9 | 2265 | 63 | |
BC113203.1 | - | 7 - 8 | 1841 | 63 |