Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smtn
Entrez Gene ID
29856
Gene Name
smoothelin
Gene Aliases
smsmo
Chromosome Location
Chr.11: 3517522 - 3539654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159284.1 NP_001152756.1 4 - 5 488 64
NM_001284427.1 NP_001271356.1 3 - 4 437 64
NM_001284428.1 NP_001271357.1 4 - 5 545 64
NM_001284429.1 NP_001271358.1 3 - 4 437 64
NM_013870.3 NP_038898.2 3 - 4 437 64
GenBank mRNA AF064236.1 - 3 - 4 384 64
AJ010305.1 - 3 - 4 384 64
AK030932.1 - 3 - 4 438 64
AK085412.1 - 3 - 4 399 64
AK138898.1 - 3 - 4 438 64
AK166385.1 - 4 - 5 505 64
AK170516.1 - 3 - 4 397 64
BC010599.1 - 3 - 4 378 64
BC066192.1 - 4 - 5 366 64
BC069836.1 - 4 - 5 366 64
BC069839.1 - 4 - 5 366 64
BY724924.1 - 3 - 4 389 64

View Full Product Details