The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013876.3 | NP_038904.1 | 2 - 3 | 563 | 74 |
GenBank mRNA | AB024427.1 | - | 3 - 4 | 428 | 74 |
AF220206.1 | - | 2 - 3 | 317 | 74 | |
AK014254.1 | - | 2 - 3 | 717 | 74 | |
AK183757.1 | - | 1 - 2 | 178 | 74 | |
AK210997.1 | - | 1 - 2 | 178 | 74 | |
BC010299.1 | - | 2 - 3 | 563 | 74 | |
BC028255.1 | - | 2 - 3 | 744 | 74 |