The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113486.1 | NP_001106958.1 | 4 - 5 | 999 | 101 |
NM_001113487.1 | NP_001106959.1 | 3 - 4 | 1179 | 101 | |
NM_001113488.1 | NP_001106960.1 | 2 - 3 | 348 | 101 | |
NM_017380.2 | NP_059076.1 | 2 - 3 | 329 | 101 | |
GenBank mRNA | AJ250723.1 | - | 2 - 3 | 335 | 101 |
AK031757.1 | - | 2 - 3 | 224 | 101 | |
AK122415.2 | - | 3 - 4 | 973 | 101 | |
AK141312.1 | - | 4 - 5 | 940 | 101 | |
AK164581.1 | - | 2 - 3 | 234 | 101 | |
BC046524.1 | - | 4 - 5 | 999 | 101 | |
BC094455.1 | - | 5 - 6 | 703 | 101 | |
BU612221.1 | - | 2 - 3 | 331 | 101 | |
BY772945.1 | - | 2 - 3 | 316 | 101 |