Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpn2
Entrez Gene ID
20014
Gene Name
ribophorin II
Gene Aliases
1300012C06Rik, AV261018, Rpn-2
Chromosome Location
Chr.2: 157279011 - 157326318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019642.4 NP_062616.2 10 - 11 1410 75
GenBank mRNA AK004968.1 - 10 - 11 1411 75
AK018761.2 - 10 - 11 1450 75
AK087923.1 - 4 - 5 447 75
AK153952.1 - 10 - 11 1271 75
AK161624.1 - 10 - 11 1221 75
AK166778.1 - 10 - 11 1196 75
AK167864.1 - 10 - 11 1190 75
AK167866.1 - 10 - 11 1190 75
AK168221.1 - 10 - 11 1190 75
BC010509.1 - 9 - 10 1183 75
BC046806.1 - 10 - 11 1438 75
D31717.1 - 9 - 10 1196 75

View Full Product Details