Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpn2
Entrez Gene ID
20014
Gene Name
ribophorin II
Gene Aliases
1300012C06Rik, AV261018, Rpn-2
Chromosome Location
Chr.2: 157279011 - 157326318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019642.4 NP_062616.2 11 - 12 1520 90
GenBank mRNA AK004968.1 - 11 - 12 1521 90
AK018761.2 - 11 - 12 1560 90
AK087923.1 - 5 - 6 89
AK153952.1 - 11 - 12 1381 90
AK161624.1 - 11 - 12 1331 90
AK166778.1 - 11 - 12 1306 90
AK167864.1 - 11 - 12 1300 90
AK167866.1 - 11 - 12 1300 90
AK168221.1 - 11 - 12 1300 90
BC010509.1 - 10 - 11 1293 90
BC046806.1 - 11 - 12 1548 90
D31717.1 - 10 - 11 1306 90

View Full Product Details