The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022411.3 | NP_071856.1 | 10 - 11 | 1473 | 67 |
GenBank mRNA | AF201903.1 | - | 10 - 11 | 1473 | 67 |
AK140972.1 | - | 10 - 11 | 1524 | 67 | |
BC013493.1 | - | 10 - 11 | 1498 | 67 |