The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023305.3 | NP_075794.3 | 4 - 5 | 1330 | 69 |
GenBank mRNA | AF275549.3 | - | 4 - 5 | 1289 | 69 |
AK028477.1 | - | 4 - 5 | 1289 | 69 | |
AK028670.1 | - | 4 - 5 | 1281 | 69 | |
AK030370.1 | - | 4 - 5 | 1271 | 69 | |
AK033121.1 | - | 4 - 5 | 1282 | 69 | |
AK051346.1 | - | - | 511 | 69 | |
AK076078.1 | - | 4 - 5 | 1289 | 69 | |
BC065996.1 | - | 4 - 5 | 1278 | 69 |