Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl37
Entrez Gene ID
56280
Gene Name
mitochondrial ribosomal protein L37
Gene Aliases
2300004O14Rik, AI132596, MRP-L2, Rpml2
Chromosome Location
Chr.4: 107055874 - 107066866 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025500.2 NP_079776.1 1 - 2 416 101
GenBank mRNA AK003811.1 - 1 - 2 408 101
AK006133.1 - 1 - 2 407 101
AK009524.1 - 1 - 2 406 101
AK161818.1 - 1 - 2 400 101
AK167461.1 - 1 - 2 409 101
AK185934.1 - 1 - 2 186 101
BC011065.1 - 1 - 2 384 101
BC028257.1 - 1 - 2 406 101

View Full Product Details