Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl37
Entrez Gene ID
56280
Gene Name
mitochondrial ribosomal protein L37
Gene Aliases
2300004O14Rik, AI132596, MRP-L2, Rpml2
Chromosome Location
Chr.4: 107055874 - 107066866 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025500.2 NP_079776.1 3 - 4 720 68
GenBank mRNA AK003811.1 - 3 - 4 712 68
AK006133.1 - 3 - 4 711 68
AK009030.1 - 3 - 4 724 68
AK009524.1 - 3 - 4 708 68
AK161818.1 - 3 - 4 704 68
AK167461.1 - 3 - 4 713 68
BC011065.1 - 3 - 4 688 68
BC028257.1 - 3 - 4 710 68

View Full Product Details