The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252384.1 | NP_001239313.1 | 7 - 8 | 1204 | 98 |
NM_001252394.1 | NP_001239323.1 | 5 - 6 | 857 | 98 | |
NM_001252395.1 | NP_001239324.1 | 7 - 8 | 1216 | 98 | |
NM_001252396.1 | NP_001239325.1 | 5 - 6 | 871 | 98 | |
NM_026071.3 | NP_080347.1 | 6 - 7 | 1041 | 98 | |
GenBank mRNA | AK005710.1 | - | 6 - 7 | 886 | 98 |
AK168145.1 | - | 5 - 6 | 656 | 98 | |
AK172622.1 | - | 5 - 6 | 843 | 98 | |
BC018167.1 | - | 6 - 7 | 847 | 98 | |
BC046767.1 | - | 6 - 7 | 891 | 98 |