Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlph
Entrez Gene ID
171531
Gene Name
melanophilin
Gene Aliases
2210418F23Rik, 5031433I09Rik, AW228792, D1Wsu84e, Slac-2a, l(1)-3Rk, l1Rk3, ln
Chromosome Location
Chr.1: 90915022 - 90951142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053015.3 NP_443748.2 1 - 2 150 58
GenBank mRNA AF384098.1 - 1 - 2 147 58
AK008973.1 - 1 - 2 122 58
AK019883.1 - 1 - 2 122 58
AK148274.1 - 1 - 2 122 58
BC006900.1 - 1 - 2 143 58
BC150804.1 - 1 - 2 44 58

View Full Product Details