Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Vmn1r52
- Entrez Gene ID
-
113849
- Gene Name
- vomeronasal 1 receptor 52
- Gene Aliases
-
V1ra7, VN3
- Chromosome Location
-
Chr.6: 90178716 - 90179645 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053222.1 |
NP_444452.1 |
1 - 1 |
550 |
145 |
- Gene Symbol
- V1ra8
- Entrez Gene ID
-
113850
- Gene Name
- vomeronasal 1 receptor, A8
- Gene Aliases
-
Vmn1r-ps33
- Chromosome Location
-
Chr.6: 90202817 - 90203656 on Build GRCm38
View Full Product Details