The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138648.2 | NP_619589.2 | 5 - 6 | 755 | 64 |
GenBank mRNA | AF303744.1 | - | 5 - 6 | 744 | 64 |
AK154687.1 | - | 5 - 6 | 756 | 64 | |
AK160919.1 | - | 5 - 6 | 755 | 64 |