The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145099.2 | NP_659567.2 | 13 - 14 | 1812 | 103 |
GenBank mRNA | AF510316.1 | - | 13 - 14 | 1812 | 103 |
BC108984.1 | - | 13 - 14 | 1798 | 103 |