The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110214.1 | NP_001103684.1 | 1 - 2 | 192 | 85 |
NM_146114.3 | NP_666226.2 | 1 - 2 | 192 | 85 | |
NM_175683.4 | NP_783614.1 | 1 - 2 | 192 | 85 | |
GenBank mRNA | AF387731.1 | - | 1 - 2 | 174 | 85 |
AK038788.1 | - | 1 - 2 | 122 | 85 | |
AK052369.1 | - | 1 - 2 | 148 | 85 | |
AK052490.1 | - | 1 - 2 | 131 | 85 | |
AK088810.1 | - | 1 - 2 | 160 | 85 | |
AK166324.1 | - | 1 - 2 | 135 | 85 | |
BC108935.2 | - | 1 - 2 | 145 | 85 | |
BY010590.1 | - | 1 - 2 | 194 | 85 | |
BY729142.1 | - | 1 - 2 | 175 | 85 |