The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110214.1 | NP_001103684.1 | 2 - 3 | 246 | 122 |
NM_146114.3 | NP_666226.2 | 2 - 3 | 246 | 122 | |
NM_175683.4 | NP_783614.1 | 2 - 3 | 246 | 122 | |
GenBank mRNA | AF387731.1 | - | 2 - 3 | 228 | 122 |
AK037126.1 | - | 2 - 3 | 213 | 122 | |
AK038788.1 | - | 2 - 3 | 176 | 122 | |
AK052369.1 | - | 2 - 3 | 202 | 122 | |
AK052490.1 | - | 2 - 3 | 185 | 122 | |
AK166324.1 | - | 2 - 3 | 189 | 122 | |
BC108935.2 | - | 2 - 3 | 199 | 122 | |
BY010590.1 | - | 2 - 3 | 248 | 122 | |
BY729142.1 | - | 2 - 3 | 229 | 122 |