The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282071.2 | NP_001269000.1 | 9 - 10 | 1014 | 107 |
NM_147779.2 | NP_680088.1 | 9 - 10 | 1083 | 107 |