The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301211.1 | NP_001288140.1 | 21 - 22 | 2647 | 74 |
NM_009677.6 | NP_033807.2 | 22 - 23 | 2656 | 74 | |
GenBank mRNA | AK028360.1 | - | 10 - 11 | 1062 | 74 |
AK034198.1 | - | 19 - 20 | 1885 | 74 | |
AK036387.1 | - | 22 - 23 | 2599 | 74 | |
AK145821.1 | - | 21 - 22 | 2578 | 74 | |
BC006691.1 | - | 21 - 22 | 2323 | 74 | |
BC052703.1 | - | 22 - 23 | 2529 | 74 | |
BC054535.1 | - | 22 - 23 | 2529 | 74 | |
X54424.1 | - | 21 - 22 | 2569 | 74 |