The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113420.1 | NP_001106891.1 | 4 - 5 | 644 | 116 |
NM_001113421.1 | NP_001106892.1 | 5 - 6 | 1328 | 116 | |
NM_001282044.1 | NP_001268973.1 | 5 - 6 | 1311 | 116 | |
NM_001282045.1 | NP_001268974.1 | 5 - 6 | 1169 | 116 | |
NM_013928.5 | NP_038956.2 | 4 - 5 | 734 | 116 | |
GenBank mRNA | AF145716.1 | - | 4 - 5 | 840 | 116 |
AK021361.1 | - | 4 - 5 | 595 | 116 | |
AK083464.1 | - | 5 - 6 | 1154 | 116 | |
AK146930.1 | - | 5 - 6 | 1285 | 116 | |
AK168005.1 | - | 5 - 6 | 1238 | 116 | |
BC060529.1 | - | 4 - 5 | 668 | 116 | |
BC094348.1 | - | 5 - 6 | 1237 | 116 | |
EU163407.1 | - | 5 - 6 | 1328 | 116 | |
EU163408.1 | - | 4 - 5 | 644 | 116 | |
KM233715.1 | - | 4 - 5 | 590 | 116 | |
KM233716.1 | - | 4 - 5 | 595 | 116 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113419.2 | NP_001106890.1 | 8 - 9 | 1506 | 116 |
GenBank mRNA | KJ941154.1 | - | 7 - 8 | 810 | 116 |