Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clasrp
Entrez Gene ID
53609
Gene Name
CLK4-associating serine/arginine rich protein
Gene Aliases
Clasp, Sfrs16, Srsf16, Swap2
Chromosome Location
Chr.7: 19581038 - 19604517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016680.5 NP_057889.3 13 - 14 1486 73
NR_125832.1 13 - 14 1486 73
NR_125833.1 13 - 14 1470 73
NR_125834.1 13 - 14 1486 73
NR_125835.1 13 - 14 1448 73
NR_125836.1 11 - 12 1350 73
GenBank mRNA AB080582.1 - 12 - 13 1349 73
AB080583.1 - 12 - 13 1349 73
AF042799.1 - 13 - 14 1451 73
AF325456.1 - 3 - 4 1332 73
AK009334.1 - 13 - 14 1437 73
AK159443.1 - 13 - 14 1455 73
BC010728.1 - 13 - 14 1437 73
BC051916.1 - 13 - 14 1461 73
BC060092.1 - 12 - 13 3854 73
BC079543.1 - 12 - 13 3848 73
BC103781.1 - 11 - 12 1316 73
BC106129.1 - 13 - 14 1449 73

View Full Product Details