Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fscn3
Entrez Gene ID
56223
Gene Name
fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus)
Gene Aliases
-
Chromosome Location
Chr.6: 28427870 - 28439398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019569.2 NP_062515.2 5 - 6 1408 67
GenBank mRNA AF176024.2 - 5 - 6 1520 67
BC050759.1 - 5 - 6 1408 67

View Full Product Details