The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253757.1 | NP_001240686.1 | 3 - 4 | 663 | 143 |
NM_023210.4 | NP_075699.3 | 3 - 4 | 663 | 143 | |
GenBank mRNA | AB037685.1 | - | 3 - 4 | 338 | 143 |
AK012759.1 | - | 3 - 4 | 630 | 143 | |
AK049647.1 | - | 3 - 4 | 630 | 143 | |
AK076049.1 | - | 3 - 4 | 490 | 143 | |
AK088401.1 | - | 3 - 4 | 643 | 143 | |
AK147888.1 | - | 3 - 4 | 629 | 143 | |
AK159446.1 | - | 3 - 4 | 630 | 143 | |
AK168380.1 | - | 3 - 4 | 612 | 143 | |
AK172146.1 | - | 3 - 4 | 677 | 143 | |
AK193570.1 | - | 1 - 2 | 125 | 143 | |
AK199406.1 | - | 1 - 2 | 125 | 143 | |
AK204175.1 | - | 1 - 2 | 125 | 143 | |
BC005690.1 | - | 3 - 4 | 598 | 143 | |
BC080684.1 | - | 3 - 4 | 603 | 143 | |
CF952181.1 | - | 3 - 4 | 334 | 143 | |
U89345.2 | - | 3 - 4 | 624 | 143 |