The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039169.1 | NP_001034258.1 | 5 - 6 | 1062 | 123 |
NM_001039170.1 | NP_001034259.1 | 5 - 6 | 611 | 123 | |
NM_023314.3 | NP_075803.2 | 5 - 6 | 611 | 123 | |
GenBank mRNA | AF068116.1 | - | 5 - 6 | 525 | 123 |
AK017221.1 | - | 5 - 6 | 1063 | 123 | |
AK018516.1 | - | 5 - 6 | 609 | 123 | |
AK032965.1 | - | 5 - 6 | 615 | 123 | |
AK076067.1 | - | 3 - 4 | 326 | 123 | |
AK138055.1 | - | 5 - 6 | 550 | 123 | |
AK150495.1 | - | 5 - 6 | 550 | 123 | |
BC028903.1 | - | 5 - 6 | 661 | 123 | |
BC045153.1 | - | 5 - 6 | 545 | 123 | |
BC049077.1 | - | 5 - 6 | 579 | 123 |