Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm14a
Entrez Gene ID
67070
Gene Name
LSM14 homolog A (SCD6, S. cerevisiae)
Gene Aliases
2700023B17Rik, AA407828, AU017544, Tral
Chromosome Location
Chr.7: 34344637 - 34389637 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025948.2 NP_080224.1 4 - 5 604 108
GenBank mRNA AK003611.1 - 2 - 3 164 108
AK012276.1 - 3 - 4 367 108
AK186175.1 - 1 - 2 124 108
BC031521.1 - 4 - 5 604 108

View Full Product Details