Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx19
Entrez Gene ID
68159
Gene Name
syntaxin 19
Gene Aliases
A030009B12Rik
Chromosome Location
Chr.16: 62814676 - 62822722 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026588.1 NP_080864.1 1 - 2 271 106
GenBank mRNA AK020701.1 - 1 - 2 222 106
AK076337.1 - - 203 106
BC023414.1 - 1 - 2 271 106

View Full Product Details