The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030890.1 | NP_112152.1 | 2 - 3 | 556 | 129 |
GenBank mRNA | BC128489.1 | - | 3 - 4 | 225 | 129 |