Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Selm
Entrez Gene ID
114679
Gene Name
selenoprotein M
Gene Aliases
1500040L08Rik, A230103K18, Sepm
Chromosome Location
Chr.11: 3514702 - 3517351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053267.2 NP_444497.1 4 - 5 524 60
GenBank mRNA AK018791.1 - 4 - 5 360 60
AK035312.1 - 4 - 5 314 60
AK046034.1 - 4 - 5 316 60
AW049879.1 - 1 - 2 60
AY043488.1 - 4 - 5 340 60
BC019742.1 - 4 - 5 216 60
BC147332.1 - 4 - 5 434 60
BC147333.1 - 4 - 5 436 60
CK787580.1 - 4 - 5 516 60

View Full Product Details