The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133363.3 | NP_579941.2 | 4 - 5 | 297 | 75 |
GenBank mRNA | AF480442.1 | - | 4 - 5 | 375 | 75 |
BC104046.2 | - | 4 - 5 | 297 | 75 | |
BC104047.1 | - | 4 - 5 | 297 | 75 |