Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptges2
Entrez Gene ID
96979
Gene Name
prostaglandin E synthase 2
Gene Aliases
0610038H10Rik, C79137, Gbf1, Mpges2, Pges2
Chromosome Location
Chr.2: 32395890 - 32402740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133783.2 NP_598544.2 3 - 4 921 68
GenBank mRNA AK002798.1 - 3 - 4 545 68
AK050616.1 - 3 - 4 547 68
AK182234.1 - 3 - 4 280 68
BC004846.1 - 3 - 4 553 68

View Full Product Details