The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127382.2 | NP_001120854.1 | 6 - 7 | 1842 | 75 |
NM_139065.3 | NP_620704.1 | 5 - 6 | 1600 | 75 | |
NM_178446.4 | NP_848541.2 | 5 - 6 | 2042 | 75 | |
GenBank mRNA | AK035620.1 | - | 5 - 6 | 1993 | 75 |
AK040440.1 | - | 2 - 3 | 1084 | 75 | |
AK085733.1 | - | 5 - 6 | 2034 | 75 | |
AK170082.1 | - | 6 - 7 | 1797 | 75 | |
BC013481.1 | - | 5 - 6 | 1612 | 75 | |
BC034195.1 | - | 4 - 5 | 1548 | 75 | |
BC112902.1 | - | 5 - 6 | 2042 | 75 |