The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139308.2 | NP_647469.2 | 5 - 6 | 1103 | 88 |
GenBank mRNA | AK083125.1 | - | 4 - 5 | 858 | 88 |
AK143699.1 | - | 5 - 6 | 1104 | 88 | |
BC017524.1 | - | 5 - 6 | 1093 | 88 | |
BC080747.1 | - | 5 - 6 | 1074 | 88 |