The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145967.2 | NP_666079.2 | 4 - 5 | 915 | 64 |
GenBank mRNA | AK032473.1 | - | 4 - 5 | 914 | 64 |
AK137363.1 | - | 4 - 5 | 837 | 64 | |
BC027127.1 | - | 4 - 5 | 867 | 64 |