The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162868.1 | NP_001156340.1 | 8 - 9 | 2379 | 67 |
NM_001162869.1 | NP_001156341.1 | 9 - 10 | 2514 | 67 | |
NM_153140.2 | NP_694780.1 | 6 - 7 | 770 | 67 | |
GenBank mRNA | AB093257.1 | - | 9 - 10 | 1955 | 67 |
AK083658.1 | - | 8 - 9 | 2063 | 67 | |
AK085516.1 | - | 6 - 7 | 771 | 67 | |
AK128985.1 | - | 6 - 7 | 770 | 67 | |
BC023364.1 | - | 6 - 7 | 761 | 67 | |
BC037132.1 | - | 6 - 7 | 810 | 67 |