Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca8a
Entrez Gene ID
217258
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8a
Gene Aliases
-
Chromosome Location
Chr.11: 110025634 - 110097583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153145.4 NP_694785.3 20 - 21 2832 90
GenBank mRNA AB255881.1 - 19 - 20 2751 90
AB255882.1 - 20 - 21 2839 90
AF498360.1 - 19 - 20 2760 90
AK031843.1 - 1 - 2 63 90
AY732492.1 - 19 - 20 2760 90
BC060032.1 - 2 - 3 326 90

View Full Product Details