Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clrn1
Entrez Gene ID
229320
Gene Name
clarin 1
Gene Aliases
A130002D11Rik, USH3, Ush3a
Chromosome Location
Chr.3: 58844028 - 58885212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153384.3 NP_700433.1 1 - 2 421 81
GenBank mRNA AF495718.1 - 1 - 2 421 81
AK037267.1 - 1 - 2 85 81

View Full Product Details