Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb13
Entrez Gene ID
69387
Gene Name
DnaJ (Hsp40) related, subfamily B, member 13
Gene Aliases
1700014P03Rik, Tsarg
Chromosome Location
Chr.7: 100503011 - 100514815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153527.2 NP_705755.2 3 - 4 390 136
GenBank mRNA AF419292.2 - 3 - 4 535 136
AK005984.1 - 3 - 4 407 136
AK139680.1 - - 137
BC048501.1 - 3 - 4 390 136

View Full Product Details