The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153587.2 | NP_705815.1 | 14 - 15 | 1987 | 94 |
GenBank mRNA | AK030051.1 | - | 12 - 13 | 1733 | 94 |
AK032316.1 | - | 13 - 14 | 1885 | 94 | |
AK046319.1 | - | 13 - 14 | 1569 | 94 | |
AK160758.1 | - | 6 - 7 | 751 | 94 | |
AY341873.1 | - | 15 - 16 | 1976 | 94 | |
AY341881.1 | - | 13 - 14 | 1781 | 94 | |
BC035298.1 | - | 15 - 16 | 1987 | 94 |