The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153587.2 | NP_705815.1 | 17 - 18 | 2500 | 72 |
GenBank mRNA | AK030051.1 | - | 15 - 16 | 2246 | 72 |
AK032316.1 | - | 16 - 17 | 2398 | 72 | |
AK160758.1 | - | 9 - 10 | 1264 | 72 | |
AY341873.1 | - | 18 - 19 | 2489 | 72 | |
AY341881.1 | - | 16 - 17 | 2294 | 72 | |
BC035298.1 | - | 18 - 19 | 2500 | 72 | |
BC038372.1 | - | 3 - 4 | 2405 | 72 |