The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170597.4 | NP_733485.1 | 1 - 2 | 678 | 63 |
GenBank mRNA | AB046110.1 | - | 1 - 2 | 996 | 63 |
AK034501.1 | - | 1 - 2 | 441 | 63 | |
AK044192.1 | - | 1 - 2 | 678 | 63 | |
AK046245.1 | - | 1 - 2 | 465 | 63 | |
AK048589.1 | - | 1 - 2 | 472 | 63 | |
AK134713.1 | - | 1 - 2 | 449 | 63 | |
AK134861.1 | - | 1 - 2 | 739 | 63 | |
BC034115.1 | - | 1 - 2 | 366 | 63 | |
BC116790.1 | - | 1 - 2 | 571 | 63 | |
BC116792.1 | - | 1 - 2 | 571 | 63 |