Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkdc
Entrez Gene ID
19090
Gene Name
protein kinase, DNA activated, catalytic polypeptide
Gene Aliases
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
Chromosome Location
Chr.16: 15637444 - 15842252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011159.2 NP_035289.2 22 - 23 2552 129
GenBank mRNA AB007544.1 - 22 - 23 2529 129
AB011543.1 - 22 - 23 2541 129
D87521.1 - 22 - 23 2529 129

View Full Product Details