Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkdc
Entrez Gene ID
19090
Gene Name
protein kinase, DNA activated, catalytic polypeptide
Gene Aliases
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
Chromosome Location
Chr.16: 15637444 - 15842252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011159.2 NP_035289.2 37 - 38 4971 127
GenBank mRNA AB007544.1 - 37 - 38 4948 127
AB011543.1 - 37 - 38 4960 127
D87521.1 - 37 - 38 4948 127

View Full Product Details