Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papln
Entrez Gene ID
170721
Gene Name
papilin, proteoglycan-like sulfated glycoprotein
Gene Aliases
E030033C16Rik
Chromosome Location
Chr.12: 83762992 - 83794875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205343.1 NP_001192272.1 25 - 26 3545 94
NM_130887.3 NP_570957.2 24 - 25 3479 94
GenBank mRNA AF314171.1 - 24 - 25 3452 94
AK053198.1 - 25 - 26 3547 94
AK086767.1 - 24 - 25 3462 94
BC005747.1 - 6 - 7 1016 94
BC132475.1 - 24 - 25 3419 94
BC137854.1 - 24 - 25 3419 94
BC144975.1 - 25 - 26 3485 94

View Full Product Details