The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139229.4 | NP_631975.3 | 1 - 2 | 473 | 56 |
GenBank mRNA | AB041610.2 | - | 1 - 2 | 396 | 56 |
AK049957.1 | - | 1 - 2 | 421 | 56 | |
AK083734.1 | - | 1 - 2 | 415 | 56 | |
AK145515.1 | - | 1 - 2 | 426 | 56 | |
AK145727.1 | - | 1 - 2 | 417 | 56 | |
AK153617.1 | - | 1 - 2 | 417 | 56 | |
AK156186.1 | - | 1 - 2 | 416 | 56 | |
AK159569.1 | - | 1 - 2 | 414 | 56 | |
AK206403.1 | - | 1 - 2 | 108 | 56 | |
BC094654.1 | - | - | 409 | 56 | |
CA324258.1 | - | 1 - 2 | 473 | 56 |