The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139229.4 | NP_631975.3 | 2 - 3 | 686 | 68 |
GenBank mRNA | AB041610.2 | - | 2 - 3 | 609 | 68 |
AK049957.1 | - | 2 - 3 | 634 | 68 | |
AK083734.1 | - | 2 - 3 | 628 | 68 | |
AK145515.1 | - | 2 - 3 | 639 | 68 | |
AK145727.1 | - | 2 - 3 | 630 | 68 | |
AK153617.1 | - | 2 - 3 | 630 | 68 | |
AK156186.1 | - | 2 - 3 | 629 | 68 | |
AK159569.1 | - | 2 - 3 | 627 | 68 | |
AK182745.1 | - | 1 - 2 | 54 | 68 | |
BC094654.1 | - | - | 622 | 68 | |
CA324258.1 | - | 2 - 3 | 686 | 68 |