The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153150.2 | NP_694790.1 | 1 - 2 | 239 | 64 |
GenBank mRNA | AK005070.1 | - | 1 - 2 | 169 | 64 |
AK159289.1 | - | 1 - 2 | 202 | 64 | |
AK170191.1 | - | 1 - 2 | 203 | 64 | |
BC037087.1 | - | 1 - 2 | 199 | 64 | |
CJ078562.1 | - | 1 - 2 | 240 | 64 |