Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a1
Entrez Gene ID
13358
Gene Name
solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1
Gene Aliases
1300019P08Rik, 2610100G11Rik, AI194714, Ctp, Dgsj, Slc20a3
Chromosome Location
Chr.16: 17925211 - 17928219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153150.2 NP_694790.1 3 - 4 440 66
GenBank mRNA AK005070.1 - 3 - 4 370 66
AK159289.1 - 3 - 4 403 66
AK170191.1 - 3 - 4 404 66
BC037087.1 - 3 - 4 400 66

View Full Product Details