The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007955.3 | NP_031981.3 | 21 - 22 | 4213 | 71 |
GenBank mRNA | AF300701.1 | - | 20 - 21 | 3690 | 71 |
AK156112.1 | - | 21 - 22 | 4216 | 71 | |
U36488.1 | - | 17 - 18 | 3375 | 71 |