Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
H2-M3
Entrez Gene ID
14991
Gene Name
histocompatibility 2, M region locus 3
Gene Aliases
H-2M3, Hmt, M3a, R4B2
Chromosome Location
Chr.17: 37270234 - 37274485 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013819.2 NP_038847.1 7 - 8 1110 104
GenBank mRNA BC031984.1 - 7 - 8 1101 104
U18797.1 - 7 - 8 1082 104

View Full Product Details